Anti-INPP5F Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: INPP5F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-569
- 77845-568
- Antigen Symbol:
- INPP5F
- INPP5F
- Antigen Synonyms:
- INPP5F|KIAA0966|SAC2|MSTP007|MSTP047|Phosphatidylinositide phosphatase SAC2|EC 3.1.3.25|Inositol polyphosphate 5-phosphatase F|Sac domain-containing inositol phosphatase 2|Sac domain-containing phosphoinositide 4-phosphatase 2|hSAC2
- INPP5F|KIAA0966|SAC2|MSTP007|MSTP047|Phosphatidylinositide phosphatase SAC2|EC 3.1.3.25|Inositol polyphosphate 5-phosphatase F|Sac domain-containing inositol phosphatase 2|Sac domain-containing phosphoinositide 4-phosphatase 2|hSAC2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2H2
- Q9Y2H2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Peptide sequence from Human Phosphatidylinositide phosphatase SAC2 protein (187-204AA)
- Peptide sequence from Human Phosphatidylinositide phosphatase SAC2 protein (187-204AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together