Anti-UTP18 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: UTP18
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-563
- 77846-564
- Antigen Symbol:
- UTP18
- UTP18
- Antigen Synonyms:
- CGI 48|U3 small nucleolar RNA associated protein 18 homolog|U3 small nucleolar RNA-associated protein 18 homolog|UTP 18|UTP18 small subunit (SSU) processome component homolog|UTP18 small subunit processome component|UTP18_HUMAN|WD repeat containing protein 50|WD repeat domain 50|WD repeat-containing protein 50|WDR50
- CGI 48|U3 small nucleolar RNA associated protein 18 homolog|U3 small nucleolar RNA-associated protein 18 homolog|UTP 18|UTP18 small subunit (SSU) processome component homolog|UTP18 small subunit processome component|UTP18_HUMAN|WD repeat containing protein 50|WD repeat domain 50|WD repeat-containing protein 50|WDR50
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5J1
- Q9Y5J1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human U3 small nucleolar RNA-associated protein 18 homolog protein (13-152AA)
- Recombinant Human U3 small nucleolar RNA-associated protein 18 homolog protein (13-152AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together