Anti-KCNIP2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: KCNIP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-462
- 77843-461
- Antigen Symbol:
- KCNIP2
- KCNIP2
- Antigen Synonyms:
- A type potassium channel modulatory protein 2|A-type potassium channel modulatory protein 2|Cardiac voltage gated potassium channel modulatory subunit|Cardiac voltage-gated potassium channel modulatory subunit|DKFZp566L1246|KChIP 2|KChIP2|KCIP2_HUMAN|KCNIP 2|Kcnip2|Kv channel interacting protein 2|Kv channel-interacting protein 2|MGC17241|Potassium channel interacting protein 2|Potassium channel-interacting protein 2
- A type potassium channel modulatory protein 2|A-type potassium channel modulatory protein 2|Cardiac voltage gated potassium channel modulatory subunit|Cardiac voltage-gated potassium channel modulatory subunit|DKFZp566L1246|KChIP 2|KChIP2|KCIP2_HUMAN|KCNIP 2|Kcnip2|Kv channel interacting protein 2|Kv channel-interacting protein 2|MGC17241|Potassium channel interacting protein 2|Potassium channel-interacting protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NS61
- Q9NS61
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Kv channel-interacting protein 2 protein (1-90AA)
- Recombinant Human Kv channel-interacting protein 2 protein (1-90AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together