Anti-UBE2I Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-474
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:UBE2I
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P63279
- Antigen Synonyms:C358B7.1|p18|SUMO 1 protein ligase|SUMO conjugating enzyme UBC9|SUMO-conjugating enzyme UBC9|SUMO-protein ligase|SUMO1 protein ligase|UBC9|UBC9_HUMAN|UBCE9|Ubiquitin carrier protein 9|Ubiquitin carrier protein|Ubiquitin carrier protein I|Ubiquitin conjugating enzyme 9|Ubiquitin conjugating enzyme E2I (homologous to yeast UBC9)|Ubiquitin conjugating enzyme E2I (UBC9 homolog|yeast)|Ubiquitin conjugating enzyme UbcE2A|Ubiquitin like protein SUMO 1 conjugating enzyme|Ubiquitin protein ligase E2I|Ubiquitin-conjugating enzyme E2 I|Ubiquitin-protein ligase I
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human SUMO-conjugating enzyme UBC9 protein (1-157AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: UBE2I
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human