Anti-DHRS7 Rabbit Recombinant polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DHRS7
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-341
- 77846-342
- Antigen Symbol:
- DHRS7
- DHRS7
- Antigen Synonyms:
- CGI 86|CGI-86|CGI86|Dehydrogenase/reductase (SDR family) member 7|Dehydrogenase/reductase SDR family member 7|DHRS7_HUMAN|DHRS7A|retDSR4|Retinal short chain dehydrogenase/reductase 4|Retinal short-chain dehydrogenase/reductase 4|retSDR4|SDR34C1|Short chain dehydrogenase/reductase family 34C|member 1
- CGI 86|CGI-86|CGI86|Dehydrogenase/reductase (SDR family) member 7|Dehydrogenase/reductase SDR family member 7|DHRS7_HUMAN|DHRS7A|retDSR4|Retinal short chain dehydrogenase/reductase 4|Retinal short-chain dehydrogenase/reductase 4|retSDR4|SDR34C1|Short chain dehydrogenase/reductase family 34C|member 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y394
- Q9Y394
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Dehydrogenase/reductase SDR family member 7 protein (29-339AA)
- Recombinant Human Dehydrogenase/reductase SDR family member 7 protein (29-339AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together