Anti-BACE2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: BACE2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77845-735
- 77845-736
- Antigen Symbol:
- BACE2
- BACE2
- Antigen Synonyms:
- ALP 56|ALP56|Asp 1|ASP1|ASP21|Aspartic Like Protease 56kDa|Aspartic-like protease 56 kDa|Aspartyl protease 1|BACE 2|BACE2_HUMAN|Beta Secretase 2|Beta Site Amyloid Beta A4 Precursor Protein Cleaving Enzyme 2|Beta-secretase 2|Beta-site amyloid precursor protein cleaving enzyme 2|Beta-site APP cleaving enzyme 2|Down region aspartic protease|Downs Syndrome Region Aspartic Protease|DRAP|Memapsin 1|Memapsin-1|Membrane associated aspartic protease 1|Membrane-associated aspartic protease 1|Theta-secretase
- ALP 56|ALP56|Asp 1|ASP1|ASP21|Aspartic Like Protease 56kDa|Aspartic-like protease 56 kDa|Aspartyl protease 1|BACE 2|BACE2_HUMAN|Beta Secretase 2|Beta Site Amyloid Beta A4 Precursor Protein Cleaving Enzyme 2|Beta-secretase 2|Beta-site amyloid precursor protein cleaving enzyme 2|Beta-site APP cleaving enzyme 2|Down region aspartic protease|Downs Syndrome Region Aspartic Protease|DRAP|Memapsin 1|Memapsin-1|Membrane associated aspartic protease 1|Membrane-associated aspartic protease 1|Theta-secretase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5Z0
- Q9Y5Z0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Beta-secretase 2 protein (84-157AA)
- Recombinant Human Beta-secretase 2 protein (84-157AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together