Anti-APOBEC3C Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: APOBEC3C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-459
- 77843-460
- Antigen Symbol:
- APOBEC3C
- APOBEC3C
- Antigen Synonyms:
- A3C|ABC3C_HUMAN|APOBEC1 like|APOBEC1-like|APOBEC1L|APOBEC3C|Apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3C|ARDC2|ARDC4|ARP5|bK150C2.3|DNA dC >dU editing enzyme APOBEC 3C|MGC19485|PBI|Phorbolin I|Phorbolin I protein|Probable DNA dC dU editing enzyme APOBEC 3C|Probable DNA dC->dU-editing enzyme APOBEC-3C
- A3C|ABC3C_HUMAN|APOBEC1 like|APOBEC1-like|APOBEC1L|APOBEC3C|Apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3C|ARDC2|ARDC4|ARP5|bK150C2.3|DNA dC >dU editing enzyme APOBEC 3C|MGC19485|PBI|Phorbolin I|Phorbolin I protein|Probable DNA dC dU editing enzyme APOBEC 3C|Probable DNA dC->dU-editing enzyme APOBEC-3C
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NRW3
- Q9NRW3
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human DNA dC->dU-editing enzyme APOBEC-3C protein (1-190AA)
- Recombinant Human DNA dC->dU-editing enzyme APOBEC-3C protein (1-190AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together