Anti-AKR1C2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77850-979
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AKR1C2
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P52895
- Antigen Synonyms:2-dihydrobenzene-1|2-diol dehydrogenase|3 alpha HSD3|3 alpha hydroxysteroid dehydrogenase type III|3-alpha-HSD3|AK1C2_HUMAN|AKR1C pseudo|Aldo keto reductase family 1 member C2|Aldo-keto reductase family 1 member C2|BABP|Bile acid binding protein|Chlordecone reductase homolog|Chlordecone reductase homolog HAKRD|DD|DD-2|DD/BABP|DD2|DDH 2|DDH2|Dihydrodiol dehydrogenase 2|Dihydrodiol dehydrogenase/bile acid binding protein|Dihydrodiol dehydrogenase/bile acid-binding protein|FLJ53800|HAKRD|HBAB|MCDR 2|MCDR2|OTTHUMP00000044759|Pseudo chlordecone reductase|SRXY8|Trans 1 2 dihydrobenzene 1 2 diol dehydrogenase|Trans-1|Type II dihydrodiol dehydrogenase|Type III 3 alpha hydroxysteroid dehydrogenase|Type III 3-alpha-hydroxysteroid dehydrogenase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Aldo-keto reductase family 1 member C2 protein (1-323AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: AKR1C2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human