Anti-HSPA12A Rabbit Recombinant Polyclonal Antibody
77857-702
New Product
- Antigen Symbol:HSPA12A
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:O43301
- Antigen Synonyms:FLJ13874|Heat shock 70 kDa protein 12A|heat shock 70kD protein 12A|heat shock 70kDa protein 12A|HS12A_HUMAN|KIAA0417
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Heat shock 70 kDa protein 12A protein (2-231AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000
Type:
Antigen: HSPA12A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human