Anti-PPP1R2 Rabbit Polyclonal Antibody
Catalog # 77848-830
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:PPP1R2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:100 µl
- Form:Liquid
- Gene ID:P41236
- Antigen Synonyms:IPP 2|IPP-2|IPP2|IPP2_HUMAN|MGC87148|Phosphatase inhibitor 2|Phosphoprotein phosphatase|PPP1R2 protein|Protein phosphatase 1 regulatory (inhibitor) subunit 2|Protein phosphatase 1 regulatory subunit 2|Protein phosphatase inhibitor 2
- Modification:phospho-Ser120/Ser121
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Peptide sequence around phosphorylation site of serine 120 and serine 121 (Q-E-S(p)-S(p)-GE) derived from Human PPP1R2.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:50 to 1:100
Type:
Antigen: PPP1R2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat