Anti-CEACAM3 Rabbit Recombinant Polyclonal Antibody
77852-939
New Product
- Antigen Symbol:CEACAM3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P40198
- Antigen Synonyms:carcinoembryonic|Carcinoembryonic antigen CGM1|Carcinoembryonic antigen gene family member 1|carcinoembryonic antigen related cell adhesion molecule 3|Carcinoembryonic antigen-related cell adhesion molecule 3|cd66 d|CD66d|CD66d antigen|CEA|CEACAM 3|CEACAM-3|CEAM 3|CEAM-3|CEAM3|CEAM3_HUMAN|CGM1|Nonspecific cross reacting antigen|W264|W282
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Carcinoembryonic antigen-related cell adhesion molecule 3 protein (1-135AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CEACAM3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human