Anti-DNAJC7 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:1000 to 1:5000, WB:1:500 to 1:2000, IHC:1:25 to 1:100
Type:
Antigen: DNAJC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77848-859
- 77848-860
- Antigen Symbol:
- DNAJC7
- DNAJC7
- Antigen Synonyms:
- DJ11|DJC7|DnaJ (Hsp40) homolog subfamily C member 7|DnaJ homolog subfamily C member 7|Dnajc7|DNJC7_HUMAN|Tetratricopeptide repeat domain 2|Tetratricopeptide repeat protein 2|TPR repeat protein 2|TPR2|TTC2
- DJ11|DJC7|DnaJ (Hsp40) homolog subfamily C member 7|DnaJ homolog subfamily C member 7|Dnajc7|DNJC7_HUMAN|Tetratricopeptide repeat domain 2|Tetratricopeptide repeat protein 2|TPR repeat protein 2|TPR2|TTC2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99615
- Q99615
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human DNAJC7
- Fusion protein of Human DNAJC7
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together