Anti-ATOX1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-554
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATOX1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O00244
- Antigen Synonyms:ATOX1|ATOX1_HUMAN|ATX1|ATX1 antioxidant protein 1 homolog (yeast)|ATX1 antioxidant protein 1 homolog|Copper transport protein|Copper transport protein ATOX1|HAH1|Metal transport protein|Metal transport protein ATX1|MGC138453|MGC138455
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Copper transport protein ATOX1 protein (1-68AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ATOX1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human