Anti-GNGT2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:200 to 1:500
Type:
Antigen: GNGT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-613
- 77856-236
- Antigen Symbol:
- GNGT2
- GNGT2
- Antigen Synonyms:
- GNG8|GNG9|GNGT8Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-T2|G gamma-C|G-gamma-8|G-gamma-9|Guanine nucleotide binding protein gamma transducing activity polypeptide 2
- GNG8|GNG9|GNGT8Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-T2|G gamma-C|G-gamma-8|G-gamma-9|Guanine nucleotide binding protein gamma transducing activity polypeptide 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14610
- O14610
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant human Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-T2 protein (1-66AA)
- Recombinant human Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-T2 protein (1-66AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together