Anti-NUP107 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NUP107
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77745-198
- 77745-199
- Antigen Symbol:
- NUP107
- NUP107
- Antigen Synonyms:
- 107 kDa nucleoporin|NU107_HUMAN|Nuclear pore complex protein Nup107|Nucleoporin 107kDa|Nucleoporin Nup107|NUP107|NUP84|yeast|homolog of|OTTHUMP00000240604|OTTHUMP00000240606
- 107 kDa nucleoporin|NU107_HUMAN|Nuclear pore complex protein Nup107|Nucleoporin 107kDa|Nucleoporin Nup107|NUP107|NUP84|yeast|homolog of|OTTHUMP00000240604|OTTHUMP00000240606
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P57740
- P57740
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nuclear pore complex protein Nup107 protein (1-66AA)
- Nuclear pore complex protein Nup107 protein (1-66AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together