Anti-MAD1L1 Rabbit Polyclonal Antibody
Catalog # 77742-533
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MAD1L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9Y6D9
- Antigen Synonyms:hMAD1|HsMAD1|MAD1|MAD1 mitotic arrest deficient like 1 (yeast)|MAD1-like protein 1|MD1L1_HUMAN|Mitotic arrest deficient 1|Mitotic arrest deficient 1-like protein 1|Mitotic checkpoint MAD1 protein homolog|Mitotic spindle assembly checkpoint protein MAD1|PIG9|Tax binding protein 181|Tax-binding protein 181|TP53I9|Tumor protein p53 inducible protein 9|TXBP181
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:TXBP181
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MAD1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat