Anti-Lgals3bp Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Lgals3bp
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Specifications
- Catalog No:
- 77858-921
- 77742-102
- Antigen Symbol:
- Lgals3bp
- Lgals3bp
- Antigen Synonyms:
- Lgals3bpGalectin-3-binding protein|Cyp-C-associated protein|CyCAP|Lectin galactoside-binding soluble 3-binding protein|Mac-2-binding protein|MAC2BP|Mac-2 BP|Protein 90K
- Lgals3bpGalectin-3-binding protein|Cyp-C-associated protein|CyCAP|Lectin galactoside-binding soluble 3-binding protein|Mac-2-binding protein|MAC2BP|Mac-2 BP|Protein 90K
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Rat
- Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O70513
- O70513
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant rat Galectin-3-binding protein (19-574AA)
- Recombinant rat Galectin-3-binding protein (19-574AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together