Anti-MYBBP1A Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA:1:2000 to 1:5000, IHC:1:50 to 1:200
Type:
Antigen: MYBBP1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77859-795
- 77743-176
- Antigen Symbol:
- MYBBP1A
- MYBBP1A
- Antigen Synonyms:
- AL024407| AU019902| cb486| FLJ37886| MBB1A_HUMAN| MYB binding protein (P160) 1a | myb binding protein (P160) 1a like| Myb-binding protein 1A| Mybbp1a| nuclear protein P160| P160| p160MBP| p53 activated protein 2| p67MBP| PAP2| PAR interacting protein| RP23 48A2.3| sb:cb486
- AL024407|AU019902|cb486|FLJ37886|MBB1A_HUMAN|MYB binding protein (P160) 1a |myb binding protein (P160) 1a like|Myb-binding protein 1A|Mybbp1a|nuclear protein P160|P160|p160MBP|p53 activated protein 2|p67MBP|PAP2|PAR interacting protein|RP23 48A2.3|sb:cb486
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BQG0
- Q9BQG0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human MYBBP1A
- Synthetic peptide of Human MYBBP1A
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together