Anti-MARK1 Rabbit Polyclonal Antibody
Catalog # 77742-784
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MARK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9P0L2
- Antigen Synonyms:KIAA1477|MAP/microtubule affinity regulating kinase 1|MAP/microtubule affinity-regulating kinase 1|MARK 1|MARK|MARK1_HUMAN|MGC126512|MGC126513|Serine/threonine protein kinase Serine/threonine-protein kinase MARK1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Serine/threonine-protein kinase MARK1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MARK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat