Anti-GNG5 Rabbit Polyclonal Antibody
Catalog # 77737-530
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GNG5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:P63218
- Antigen Synonyms:FLJ92393|GBG5_HUMAN|Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-5|OTTHUMP00000011474|OTTHUMP00000011565
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:FLJ92393
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IF: 1:200 to 1:1000, ELISA: 1:20000
Type:
Antigen: GNG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat