Anti-MYL1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MYL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-746
- 77860-209
- Antigen Symbol:
- MYL1
- MYL1
- Antigen Synonyms:
- A1 catalytic|Alkali myosin light chain 1|MLC1/MLC3|MLC1F|MLC1F/MLC3F|MLC3F|Myl1|MYL1_HUMAN|Myosin light chain 1 skeletal muscle isoform|Myosin light chain 1/3|Myosin light chain 1/3, skeletal muscle isoform|Myosin light chain A1/A2|Myosin light chain alkali 1/2|skeletal muscle isoform
- A1 catalytic|Alkali myosin light chain 1|MLC1/MLC3|MLC1F|MLC1F/MLC3F|MLC3F|Myl1|MYL1_HUMAN|Myosin light chain 1 skeletal muscle isoform|Myosin light chain 1/3|Myosin light chain 1/3, skeletal muscle isoform|Myosin light chain A1/A2|Myosin light chain alkali 1/2|skeletal muscle isoform
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P05976
- P05976
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Myosin light chain 1/3, skeletal muscle isoform protein (1-150AA)
- Recombinant Human Myosin light chain 1/3, skeletal muscle isoform protein (1-150AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together