Anti-EMCN Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: EMCN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77854-692
- 77735-841
- Antigen Symbol:
- EMCN
- Antigen Synonyms:
- EMCN|EMCN2|Endomucin 2|Endomucin|Endomucin-2|Endothelial sialomucin|Gastric cancer antigen Ga34|MUC-14|MUC14|MUCEN_HUMAN|Mucin 14|Mucin-14
- EMCN|EMCN2|Endomucin 2|Endomucin|Endomucin-2|Endothelial sialomucin|Gastric cancer antigen Ga34|MUC-14|MUC14|MUCEN_HUMAN|Mucin 14|Mucin-14
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- Q9ULC0
- Q9ULC0
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Endomucin protein (19-135AA)
- Recombinant Human Endomucin protein (19-135AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together