Anti-HRH2 Rabbit Polyclonal Antibody
Catalog # 77738-008
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HRH2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P25021
- Antigen Synonyms:Gastric receptor 1|Gastric receptor I|H2R|HGNC:5183|HH2R|Histamine H2 receptor|Histamine receptor H2|HRH2_HUMAN|OTTHUMP00000161242|OTTHUMP00000161243
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Gastric receptor 1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IF: 1:200 to 1:1000, ELISA: 1:5000
Type:
Antigen: HRH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human