Anti-NKX3-2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NKX3-2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-505
- 77744-506
- Antigen Symbol:
- NKX3-2
- NKX3-2
- Antigen Synonyms:
- Bagpipe homeobox 1|Bagpipe homeobox homolog 1|Bagpipe homeobox protein homolog 1|Bagpipe homeobox|Drosophila|homolog of|1|BAPX 1|Homeobox protein NK-3 homolog B|Homeobox protein Nkx 3.2|Homeobox protein Nkx-3.2|MGC138171|NK3 homeobox 2|NKX3 2|NKX3.2|mouse|NKX32_HUMAN|NKX3B|OTTHUMP00000115816|SMMD
- Bagpipe homeobox 1|Bagpipe homeobox homolog 1|Bagpipe homeobox protein homolog 1|Bagpipe homeobox|Drosophila|homolog of|1|BAPX 1|Homeobox protein NK-3 homolog B|Homeobox protein Nkx 3.2|Homeobox protein Nkx-3.2|MGC138171|NK3 homeobox 2|NKX3 2|NKX3.2|mouse|NKX32_HUMAN|NKX3B|OTTHUMP00000115816|SMMD
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P78367
- P78367
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Homeobox protein Nkx-3.2 protein (1-333AA)
- Homeobox protein Nkx-3.2 protein (1-333AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together