Order Entry
United States
ContactUsLinkComponent
Anti-Map2k4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-Map2k4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-Map2k4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77859-373
  • 77742-644
  • Antigen Symbol:
  • Map2k4
  • Map2k4
  • Antigen Synonyms:
  • Jnkk1|Mek4|Mkk4|Prkmk4|Sek1|Serk1|Skk1|Dual specificity mitogen-activated protein kinase kinase 4|MAP kinase kinase 4|MAPKK 4|EC 2.7.12.2|C-JUN N-terminal kinase kinase 1|JNK kinase 1|JNKK 1|JNK-activating kinase 1|MAPK/ERK kinase 4|MEK 4|SAPK/ERK kinase 1
  • Jnkk1|Mek4|Mkk4|Prkmk4|Sek1|Serk1|Skk1|Dual specificity mitogen-activated protein kinase kinase 4|MAP kinase kinase 4|MAPKK 4|EC 2.7.12.2|C-JUN N-terminal kinase kinase 1|JNK kinase 1|JNKK 1|JNK-activating kinase 1|MAPK/ERK kinase 4|MEK 4|SAPK/ERK kinase 1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P47809
  • P47809
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Mouse Dual specificity mitogen-activated protein kinase kinase 4 protein (2-397AA)
  • Recombinant Mouse Dual specificity mitogen-activated protein kinase kinase 4 protein (2-397AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together