Anti-GSTA4 Rabbit Recombinant polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: GSTA4
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-146
- 77856-542
- Antigen Symbol:
- GSTA4
- GSTA4
- Antigen Synonyms:
- DKFZp686D21185| Glutathione S alkyltransferase A4| Glutathione S aralkyltransferase A4| Glutathione S aryltransferase A4| Glutathione S transferase A4 4| Glutathione S transferase A4| Glutathione S transferase alpha 4| Glutathione S-transferase A4| Glutathione S-transferase A4-4| Glutathione transferase A4 4| GST class alpha member 4| GST class-alpha member 4| GSTA4 4| GSTA4_HUMAN| GTA4| OTTHUMP00000016624| OTTHUMP00000016625| S (hydroxyalkyl)glutathione lyase A4
- DKFZp686D21185|Glutathione S alkyltransferase A4|Glutathione S aralkyltransferase A4|Glutathione S aryltransferase A4|Glutathione S transferase A4 4|Glutathione S transferase A4|Glutathione S transferase alpha 4|Glutathione S-transferase A4|Glutathione S-transferase A4-4|Glutathione transferase A4 4|GST class alpha member 4|GST class-alpha member 4|GSTA4 4|GSTA4_HUMAN|GTA4|OTTHUMP00000016624|OTTHUMP00000016625|S (hydroxyalkyl)glutathione lyase A4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15217
- O15217
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Glutathione S-transferase A4 protein (1-222AA)
- Recombinant Human Glutathione S-transferase A4 protein (1-222AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together