Anti-Mmp7 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77743-259
- 77859-853
- Antigen Symbol:
- Mmp7
- Mmp7
- Antigen Synonyms:
- Mmp-7Matrilysin|EC 3.4.24.23|Matrin|Matrix metalloproteinase-7|MMP-7|Pump-1 protease|Uterine metalloproteinase
- Mmp-7Matrilysin|EC 3.4.24.23|Matrin|Matrix metalloproteinase-7|MMP-7|Pump-1 protease|Uterine metalloproteinase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Rat
- Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P50280
- P50280
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Rat Matrilysin protein (98-267AA)
- Recombinant Rat Matrilysin protein (98-267AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together