Anti-EXOSC2 Rabbit Polyclonal Antibody
77736-298
New Product
- Antigen Symbol:EXOSC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q13868
- Antigen Synonyms:RRP4Exosome complex component RRP4|Exosome component 2|Ribosomal RNA-processing protein 4
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Ribosomal RNA-processing protein 4
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human