Anti-MDK Rabbit Polyclonal Antibody
Catalog # 77742-930
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MDK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:100 µl
- Form:Liquid
- Gene ID:P21741
- Antigen Synonyms:Amphiregulin associated protein|Amphiregulin-associated protein|ARAP|FLJ27379|Mdk|Midgestation and kidney protein|Midkine|MK 1|MK|MK_HUMAN|MK1|NEGF 2|NEGF2|Neurite growth promoting factor 2|Neurite outgrowth promoting protein|Neurite outgrowth-promoting factor 2|Neurite outgrowth-promoting protein|Retinoic acid inducible factor
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Retinoic acid inducible factor
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MDK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat