Anti-Nkx2-2 Rabbit Recombinant Polyclonal Antibody
New Product
Type:
Antigen: Nkx2-2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
- Catalog No:
- 77744-477
- 77744-478
- Antigen Symbol:
- Nkx2-2
- Nkx2-2
- Antigen Synonyms:
- Nkx-2.2|Nkx2b|Homeobox protein Nkx-2.2|Homeobox protein NK-2 homolog B
- Nkx-2.2|Nkx2b|Homeobox protein Nkx-2.2|Homeobox protein NK-2 homolog B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P42586
- P42586
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Homeobox protein Nkx-2.2 protein (1-273AA)
- Homeobox protein Nkx-2.2 protein (1-273AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together