Anti-FANCA Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: FANCA
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-839
- 77855-192
- Antigen Symbol:
- FANCA
- FANCA
- Antigen Synonyms:
- FA 1| FA| FA H| FA1| FAA| FACA| FAH| FANCA_HUMAN| FANCH| Fanconi anemia complementation group A| Fanconi anemia complementation group H| Fanconi anemia group A protein| Fanconi anemia type 1| MGC75158| Protein FACA
- FA 1|FA|FA H|FA1|FAA|FACA|FAH|FANCA_HUMAN|FANCH|Fanconi anemia complementation group A|Fanconi anemia complementation group H|Fanconi anemia group A protein|Fanconi anemia type 1|MGC75158|Protein FACA
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15360
- O15360
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Fanconi anemia group A protein (609-813AA)
- Recombinant Human Fanconi anemia group A protein (609-813AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together