Anti-MAN1A2 Rabbit Polyclonal Antibody
Catalog # 77742-607
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MAN1A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O60476
- Antigen Synonyms:Processing alpha-1,2-mannosidase IB|EC 3.2.1.113|MAN1BMannosyl-oligosaccharide 1,2-alpha-mannosidase IB|Mannosidase alpha class 1A member 2|Alpha-1,2-mannosidase IB
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Alpha-1,2-mannosidase IB
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MAN1A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat