Anti-LGI1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: LGI1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-935
- 77742-119
- Antigen Symbol:
- LGI1
- Antigen Synonyms:
- ADLTE|ADPAEF|ADPEAF|Epitempin 1|EPITEMPIN|Epitempin-1|EPT|ETL1|IB1099|leucine rich glioma inactivated 1|Leucine rich glioma-inactivated protein 1|Leucine-rich glioma-inactivated protein 1|LGI1|LGI1_HUMAN|OTTHUMP00000020121|OTTHUMP00000020122
- ADLTE|ADPAEF|ADPEAF|Epitempin 1|EPITEMPIN|Epitempin-1|EPT|ETL1|IB1099|leucine rich glioma inactivated 1|Leucine rich glioma-inactivated protein 1|Leucine-rich glioma-inactivated protein 1|LGI1|LGI1_HUMAN|OTTHUMP00000020121|OTTHUMP00000020122
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- O95970
- O95970
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Leucine-rich glioma-inactivated protein 1 protein (35-285AA)
- Recombinant Human Leucine-rich glioma-inactivated protein 1 protein (35-285AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together