Anti-NR1H2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NR1H2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-917
- 77744-916
- Antigen Symbol:
- NR1H2
- NR1H2
- Antigen Synonyms:
- Liver X nuclear receptor beta|Liver X receptor beta|LX receptor beta|LXR b|LXR beta|LXRB|NER|NER I|NER1|NR1H 2|NR1H2_HUMAN|Nuclear orphan receptor LXR beta|Nuclear receptor NER|Nuclear receptor subfamily 1 group H member 2|OR-1|Oxysterols receptor LXR-beta|RIP15|Steroid hormone nuclear receptor NER|Ubiquitously-expressed nuclear receptor|UNR
- Liver X nuclear receptor beta|Liver X receptor beta|LX receptor beta|LXR b|LXR beta|LXRB|NER|NER I|NER1|NR1H 2|NR1H2_HUMAN|Nuclear orphan receptor LXR beta|Nuclear receptor NER|Nuclear receptor subfamily 1 group H member 2|OR-1|Oxysterols receptor LXR-beta|RIP15|Steroid hormone nuclear receptor NER|Ubiquitously-expressed nuclear receptor|UNR
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P55055
- P55055
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Oxysterols receptor LXR-beta protein (1-461AA)
- Oxysterols receptor LXR-beta protein (1-461AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together