Anti-NQO1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NQO1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-881
- 77744-882
- Antigen Symbol:
- NQO1
- NQO1
- Antigen Synonyms:
- Azoreductase|Cytochrome b 5 reductase|DHQU|DIA 4|DIA4|Diaphorase (NADH/NADPH) (cytochrome b 5 reductase)|Diaphorase (NADH/NADPH)|Diaphorase 4|Dioxin inducible 1|DT diaphorase|DT-diaphorase|DTD|Menadione reductase|NAD(P)H dehydrogenase [quinone] 1|NAD(P)H dehydrogenase quinone 1|NAD(P)H menadione oxidoreductase 1 dioxin inducible|NAD(P)H quinone dehydrogenase 1|NAD(P)H: menadione oxidoreductase 1 dioxin inducible 1|NAD(P)H:menadione oxidoreductase 1|NAD(P)H:Quinone acceptor oxidoreductase type 1|NAD(P)H:quinone oxidoreductase 1|NAD(P)H:quinone oxireductase|NMOR 1|NMOR I|NMOR1|NMORI|NQO 1|NQO1_HUMAN|Phylloquinone reductase|QR 1|QR1|Quinone reductase 1
- Azoreductase|Cytochrome b 5 reductase|DHQU|DIA 4|DIA4|Diaphorase (NADH/NADPH) (cytochrome b 5 reductase)|Diaphorase (NADH/NADPH)|Diaphorase 4|Dioxin inducible 1|DT diaphorase|DT-diaphorase|DTD|Menadione reductase|NAD(P)H dehydrogenase [quinone] 1|NAD(P)H dehydrogenase quinone 1|NAD(P)H menadione oxidoreductase 1 dioxin inducible|NAD(P)H quinone dehydrogenase 1|NAD(P)H: menadione oxidoreductase 1 dioxin inducible 1|NAD(P)H:menadione oxidoreductase 1|NAD(P)H:Quinone acceptor oxidoreductase type 1|NAD(P)H:quinone oxidoreductase 1|NAD(P)H:quinone oxireductase|NMOR 1|NMOR I|NMOR1|NMORI|NQO 1|NQO1_HUMAN|Phylloquinone reductase|QR 1|QR1|Quinone reductase 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P15559
- P15559
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- NAD(P)H dehydrogenase [quinone] 1 protein (55-155AA)
- NAD(P)H dehydrogenase [quinone] 1 protein (55-155AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together