Anti-MMP2 Rabbit Recombinant polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:200-1:500
Type:
Antigen: MMP2
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-247
- 77859-842
- Antigen Symbol:
- MMP2
- MMP2
- Antigen Synonyms:
- Matrix metalloproteinase II|TBE-1|CLG4A|MMP-2|72 kDa gelatinase|Gelatinase neutrophil|TBE 1|MONA|CLG 4|MMP 2|Matrix Metalloproteinase 2|MMP2_HUMAN|Neutrophil gelatinase|CLG 4A|Gelatinase alpha|Matrix metallopeptidase 2 gelatinase A 72kDa gelatinase 72kDa type IV collagenase|Collagenase type IV A|Gelatinase A|PEX|Collagenase Type 4 alpha|CLG4|Matrix metalloproteinase-2|72kD type IV collagenase|Matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase)|MMP II
- 72 kDa gelatinase|72kD type IV collagenase|CLG 4|CLG 4A|CLG4|CLG4A|Collagenase Type 4 alpha|Collagenase type IV A|Gelatinase A|Gelatinase alpha|Gelatinase neutrophil|Matrix metallopeptidase 2 gelatinase A 72kDa gelatinase 72kDa type IV collagenase|Matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase)|Matrix Metalloproteinase 2|Matrix metalloproteinase II|Matrix metalloproteinase-2|MMP 2|MMP II|MMP-2|MMP2_HUMAN|MONA|Neutrophil gelatinase|PEX|TBE 1|TBE-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P08253
- P08253
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human 72 kDa type IV collagenase protein (82-292AA)
- Recombinant Human 72 kDa type IV collagenase protein (82-292AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together