Anti-MTMR14 Rabbit Polyclonal Antibody
Catalog # 77743-609
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MTMR14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8NCE2
- Antigen Synonyms:C3orf29|Egg derived tyrosine phosphatase homolog|FLJ11546|FLJ22405|FLJ46453|FLJ90311|HCV NS5A transactivated protein 4 splice variant A binding protein 1|HCV NS5A-transactivated protein 4 splice variant A-binding protein 1|hEDTP|hJumpy|jumpy|MTMR 14|MTMRE_HUMAN|Myotubularin related protein 14|Myotubularin-related protein 14|NS5ATP4ABP1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:NS5ATP4ABP1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MTMR14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat