Anti-KCNK3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: KCNK3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-518
- 77858-459
- Antigen Symbol:
- KCNK3
- KCNK3
- Antigen Synonyms:
- Acid sensitive potassium channel protein TASK 1|Acid sensitive potassium channel protein TASK|Acid-sensitive potassium channel protein TASK-1|Cardiac two pore background K(+) channel|cTBAK 1|K2p3.1|KCNK3_HUMAN|OAT1|Potassium channel subfamily K member 3|potassium channel|subfamily K|member 3|Potassium inwardly rectifying channel subfamily K member 3|PPH4|rTASK|TASK 1|TASK|TBAK1|TWIK related acid sensitive K+ channel|TWIK-related acid-sensitive K(+) channel 1|Two P domain potassium channel|Two pore K(+) channel KT3.|Two pore K(+) channel KT3.1|Two pore potassium channel KT3.1
- Acid sensitive potassium channel protein TASK 1|Acid sensitive potassium channel protein TASK|Acid-sensitive potassium channel protein TASK-1|Cardiac two pore background K(+) channel|cTBAK 1|K2p3.1|KCNK3_HUMAN|OAT1|Potassium channel subfamily K member 3|potassium channel|subfamily K|member 3|Potassium inwardly rectifying channel subfamily K member 3|PPH4|rTASK|TASK 1|TASK|TBAK1|TWIK related acid sensitive K+ channel|TWIK-related acid-sensitive K(+) channel 1|Two P domain potassium channel|Two pore K(+) channel KT3.|Two pore K(+) channel KT3.1|Two pore potassium channel KT3.1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14649
- O14649
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Potassium channel subfamily K member 3 protein (278-379AA)
- Recombinant Human Potassium channel subfamily K member 3 protein (278-379AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together