Anti-Klk7 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Klk7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Specifications
- Catalog No:
- 77858-655
- 77741-774
- Antigen Symbol:
- Klk7
- Klk7
- Antigen Synonyms:
- Klk7| Klk-7| Glandular kallikrein-7| submandibular/renal| rGK-7| EC 3.4.21.35| Esterase B| Kallikrein-related protein K1| Proteinase A| RSKG-7| Tissue kallikrein
- Klk7|Klk-7|Glandular kallikrein-7|submandibular/renal|rGK-7|EC 3.4.21.35|Esterase B|Kallikrein-related protein K1|Proteinase A|RSKG-7|Tissue kallikrein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Rat
- Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P36373
- P36373
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Rat Glandular kallikrein-7, submandibular/renal protein (25-261AA)
- Recombinant Rat Glandular kallikrein-7, submandibular/renal protein (25-261AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together