Anti-MAPK13 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAPK13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-455
- 77742-739
- Antigen Symbol:
- MAPK13
- MAPK13
- Antigen Synonyms:
- MAP kinase 13|MAP kinase p38 delta|MAPK 13|MAPK-13|MGC99536|Mitogen activated protein kinase 13|Mitogen-activated protein kinase 13|Mitogen-activated protein kinase p38 delta|MK13_HUMAN|OTTHUMP00000016282|OTTHUMP00000016283|p38 delta|P38delta|PRKM13|SAPK 4|SAPK4|Stress activated protein kinase 4|Stress-activated protein kinase 4
- MAP kinase p38 delta|Mitogen-activated protein kinase 13|SAPK 4|MAPK 13|MGC99536|PRKM13|Stress-activated protein kinase 4|MAP kinase 13|OTTHUMP00000016282|Mitogen activated protein kinase 13|OTTHUMP00000016283|MAPK-13|p38 delta|P38delta|MK13_HUMAN|SAPK4|Mitogen-activated protein kinase p38 delta|Stress activated protein kinase 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15264
- O15264
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mitogen-activated protein kinase 13 protein (1-365AA)
- Recombinant Human Mitogen-activated protein kinase 13 protein (1-365AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together