Anti-HLA-G Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: HLA-G
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-461
- 77740-110
- Antigen Symbol:
- HLA-G
- HLA-G
- Antigen Synonyms:
- B2 microglobulin|DADB-15K14.8|HLA 6.0|HLA class I histocompatibility antigen alpha chain G|HLA class I histocompatibility antigen|alpha chain G|HLA class I molecule|HLA G|HLA G antigen|HLA G histocompatibility antigen class I G|HLA G3|HLA-G histocompatibility antigen|class I|HLA60|HLAG|HLAG_HUMAN|Major histocompatibility complex class I G|MHC class I antigen|MHC class I antigen G|MHC G|T-cell A locus|TCA
- B2 microglobulin|DADB-15K14.8|HLA 6.0|HLA class I histocompatibility antigen alpha chain G|HLA class I histocompatibility antigen|alpha chain G|HLA class I molecule|HLA G|HLA G antigen|HLA G histocompatibility antigen class I G|HLA G3|HLA-G histocompatibility antigen|class I|HLA60|HLAG|HLAG_HUMAN|Major histocompatibility complex class I G|MHC class I antigen|MHC class I antigen G|MHC G|T-cell A locus|TCA
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P17693
- P17693
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human HLA class I histocompatibility antigen, alpha chain G protein (20-230AA)
- Recombinant human HLA class I histocompatibility antigen, alpha chain G protein (20-230AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together