Anti-ADGRF3 Rabbit Polyclonal Antibody
Catalog # 77735-967
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ADGRF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q8IZF5
- Antigen Synonyms:GPR113|PGR23|UNQ9196/PRO34000Adhesion G-protein coupled receptor F3|G-protein coupled receptor 113|G-protein coupled receptor PGR23
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:G-protein coupled receptor PGR23
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:40000
Type:
Antigen: ADGRF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human