Anti-PTTG1IP Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PTTG1IP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-059
- 77855-295
- Antigen Symbol:
- PTTG1IP
- PTTG1IP
- Antigen Synonyms:
- 1810010L20Rik|AI314311|AU018448|C21orf1|C21orf3|C79540|chromosome 21 open reading frame 1|MGC36923|PBF|Pituitary tumor transforming 1 interacting protein|Pituitary tumor transforming gene 1 protein interacting protein|Pituitary tumor transforming gene protein binding factor|Pituitary tumor-transforming gene 1 protein-interacting protein|Pituitary tumor-transforming gene protein-binding factor|PTTG binding factor|PTTG-binding factor|PTTG_HUMAN|Pttg1ip|Putative surface glycoprotein C21orf1 precursor
- 1810010L20Rik| AI314311| AU018448| C21orf1| C21orf3| C79540| chromosome 21 open reading frame 1| MGC36923| PBF| Pituitary tumor transforming 1 interacting protein| Pituitary tumor transforming gene 1 protein interacting protein| Pituitary tumor transforming gene protein binding factor| Pituitary tumor-transforming gene 1 protein-interacting protein| Pituitary tumor-transforming gene protein-binding factor| PTTG binding factor| PTTG-binding factor| PTTG_HUMAN| Pttg1ip| Putative surface glycoprotein C21orf1 precursor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P53801
- P53801
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Pituitary tumor-transforming gene 1 protein-interacting protein (118-180AA)
- Recombinant Human Pituitary tumor-transforming gene 1 protein-interacting protein (118-180AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together