Anti-GHITM Rabbit Polyclonal Antibody
77737-495
New Product
- Antigen Symbol:GHITM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9H3K2
- Antigen Synonyms:Dermal papilla derived protein 2|Dermal papilla-derived protein 2|DERP2|Ghitm|GHITM_HUMAN|Growth hormone inducible transmembrane protein|Growth hormone-inducible transmembrane protein|HSPC282|MICS1|Mitochondrial morphology and cristae structure 1|My021|PTD010|TMBIM5|Transmembrane BAX inhibitor motif containing 5|Transmembrane BAX inhibitor motif containing protein 5|Transmembrane BAX inhibitor motif-containing protein 5
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Dermal papilla derived protein 2
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:10000
Type:
Antigen: GHITM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human