Anti-XIAP Rabbit Polyclonal Antibody
Catalog # 77738-043
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:XIAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P98170
- Antigen Synonyms:AP 13|API3|Apoptosis Inhibitor 3|Baculoviral IAP repeat containing 4|Baculoviral IAP Repeat Containing Protein 4|Baculoviral IAP repeat-containing protein 4|BIRC 4|BIRC4|E3 ubiquitin-protein ligase hIAP-3|hIAP3|HILP|IAP 3|IAP like protein|IAP-3|IAP-like protein|IAP3|ILP 1|ILP|ILP1|Inhibitor of apoptosis protein 3|Inhibitor of Apoptosis X Linked|Mammalian IAP Homologue A|MIHA|X linked IAP|X linked inhibitor of apoptosis|X linked inhibitor of apoptosis E3 ubiquitin protein ligase|X linked inhibitor of apoptosis protein|X-linked IAP|x-linked inhibitor of apopotosis proteins|X-linked inhibitor of apoptosis protein|XIAP_HUMAN|XLP2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:AP 13
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:20000
Type:
Antigen: XIAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat