Anti-NID1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
Anti-NID1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
Certificates
About this item
Type:
Antigen: NID1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type:
Antigen: NID1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications for Product Family
Specifications
- Catalog No:
- 77744-431
- 77744-432
- Antigen Symbol:
- NID1
- NID1
- Antigen Synonyms:
- Enactin|Ent|Entactin 1|Entactin|NID 1|NID|NID-1|NID1_HUMAN|Nidogen (enactin)|Nidogen 1|Nidogen|Nidogen-1
- Enactin|Ent|Entactin 1|Entactin|NID 1|NID|NID-1|NID1_HUMAN|Nidogen (enactin)|Nidogen 1|Nidogen|Nidogen-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14543
- P14543
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nidogen-1 protein (792-942AA)
- Nidogen-1 protein (792-942AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Product Family Options
Product Information
- SizePackagingAvailabilityPrice
- Supplier Number: A29703-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77744-431Antigen SymbolNID1Antigen SynonymsEnactin|Ent|Entactin 1|Entactin|NID 1|NID|NID-1|NID1_HUMAN|Nidogen (enactin)|Nidogen 1|Nidogen|Nidogen-1ClonalityRecombinant PolyclonalConjugationHRP (Horseradish Peroxidase)ReactivityHumanHostRabbitGene IDP14543IsotypeIgGELISAYesImmunogenNidogen-1 protein (792-942AA)FormLiquidPurification>95%, protein G purifiedStorage Buffer0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.Storage TemperatureUpon receipt, store at −20 or −80 °C. Avoid repeated freeze.Shipping TemperatureShipped on blue ice - Supplier Number: A29703-50UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77744-432Antigen SymbolNID1Antigen SynonymsEnactin|Ent|Entactin 1|Entactin|NID 1|NID|NID-1|NID1_HUMAN|Nidogen (enactin)|Nidogen 1|Nidogen|Nidogen-1ClonalityRecombinant PolyclonalConjugationHRP (Horseradish Peroxidase)ReactivityHumanHostRabbitGene IDP14543IsotypeIgGELISAYesImmunogenNidogen-1 protein (792-942AA)FormLiquidPurification>95%, protein G purifiedStorage Buffer0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.Storage TemperatureUpon receipt, store at −20 or −80 °C. Avoid repeated freeze.Shipping TemperatureShipped on blue ice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



