Anti-FANCL Rabbit Polyclonal Antibody
Catalog # 77736-851
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FANCL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NW38
- Antigen Synonyms:E3 ubiquitin-protein ligase FAAP43|FANCL_HUMAN|Fanconi anemia complementation group L|Fanconi anemia group L protein|Fanconi anemia-associated polypeptide of 43 kDa|FLJ10335|PHD finger protein 9|PHF9|POG|Ubiquitin ligase protein FANCL
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Ubiquitin ligase protein FANCL
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: FANCL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat