Anti-IDO1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: IDO1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77857-821
- 77740-609
- Antigen Symbol:
- IDO1
- IDO1
- Antigen Synonyms:
- 3-dioxygenase|I23O1_HUMAN|IDO 1|IDO|IDO-1|INDO|indolamine 2,3 dioxygenase|Indole 2 3 dioxygenase|indoleamine 2 3 dioxygenase 1|indoleamine 2 3 dioxygenase|Indoleamine 2,3-dioxygenase 1|Indoleamine pyrrole 2 3 dioxygenase|Indoleamine-pyrrole 2
- 3-dioxygenase|I23O1_HUMAN|IDO 1|IDO|IDO-1|INDO|indolamine 2,3 dioxygenase|Indole 2 3 dioxygenase|indoleamine 2 3 dioxygenase 1|indoleamine 2 3 dioxygenase|Indoleamine 2,3-dioxygenase 1|Indoleamine pyrrole 2 3 dioxygenase|Indoleamine-pyrrole 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14902
- P14902
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Indoleamine 2,3-dioxygenase 1 protein (1-403AA)
- Recombinant human Indoleamine 2,3-dioxygenase 1 protein (1-403AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together