Order Entry
United States
ContactUsLinkComponent
Anti-GHR Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-GHR Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-GHR Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77738-236
  • 77855-979
  • Antigen Symbol:
  • GHR
  • GHR
  • Antigen Synonyms:
  • GH receptor|GH-binding protein|GHBP|GHBP, included|GHR|GHR_HUMAN|Growth hormone binding protein|Growth hormone receptor|Growth hormone receptor precursor|Growth hormone-binding protein|Growth hormone-binding protein, included|Increased responsiveness to growth hormone, included|Serum binding protein|Serum-binding protein|Somatotropin receptor
  • GH receptor|GH-binding protein|GHBP|GHBP, included|GHR|GHR_HUMAN|Growth hormone binding protein|Growth hormone receptor|Growth hormone receptor precursor|Growth hormone-binding protein|Growth hormone-binding protein, included|Increased responsiveness to growth hormone, included|Serum binding protein|Serum-binding protein|Somatotropin receptor
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P10912
  • P10912
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Growth hormone receptor protein (19-184AA)
  • Recombinant Human Growth hormone receptor protein (19-184AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together