Anti-ETS1 Rabbit Polyclonal Antibody
Catalog # 77736-598
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ETS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:P14921
- Antigen Synonyms:Avian erythroblastosis virus E26 (v ets) oncogene homolog 1|C ets 1 protein|c-ets-1|ETS 1|Ets protein|ETS proto-oncogene 1|transcription factor|ETS1|ETS1 oncogene|ETS1 protein|ETS1_HUMAN|EWSR 2|EWSR2|FLJ10768|Oncogene ETS1|P54|Protein C-ets-1|v ets avian erythroblastosis virus E2 oncogene homolog|v ets avian erythroblastosis virus E2 oncogene homolog 1|v ets avian erythroblastosis virus E26 oncogene homolog 1|v ets erythroblastosis virus E26 oncogene homolog 1|v-ets erythroblastosis virus E26 oncogene homolog 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Avian erythroblastosis virus E26 (v ets) oncogene homolog 1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: ETS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat